missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) 2,4-pentanedionate, Thermo Scientific Chemicals

$161.88 - $448.38

Chemical Identifiers

CAS 17501-44-9
Molecular Formula C20H28O8Zr
Molecular Weight (g/mol) 487.66
MDL Number MFCD00000036
InChI Key DFPGFWYWSAYLCW-UHFFFAOYSA-N
Synonym 4-hydroxypent-3-en-2-one; zirconium
PubChem CID 50912253
IUPAC Name 4-hydroxypent-3-en-2-one;zirconium
SMILES [Zr+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1406518
View Documents
Thermo Scientific Chemicals
01406518
50 g
Each for $161.88
Only null left
 
AA1406530
View Documents
Thermo Scientific Chemicals
01406530
250 g
Each for $448.38
Only null left
 
Description

Description

Zirconium(IV) 2,4-pentanedionate is used as a cross-linking agent for oxygen-containing polymers. It is used as a catalyst in organic synthesis for the nitration of o-nitrotoluene using a dinitrogen pentoxide to get 2,4-dinitrotoluenes and 2,6-dinitrotoluenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17501-44-9
487.66
DFPGFWYWSAYLCW-UHFFFAOYSA-N
50912253
[Zr+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
C20H28O8Zr
MFCD00000036
4-hydroxypent-3-en-2-one; zirconium
4-hydroxypent-3-en-2-one;zirconium

Specifications

17501-44-9
White
C20H28O8Zr
50 g
Soluble in water.
[Zr+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
487.66
487.66
Zirconium(IV) 2,4-pentanedionate
191°C to 195°C
Weak
MFCD00000036
4-hydroxypent-3-en-2-one; zirconium
DFPGFWYWSAYLCW-UHFFFAOYSA-N
4-hydroxypent-3-en-2-one;zirconium
50912253
Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 241-510-5

RTECSNumber : ZH9280000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only