accessibility menu, dialog, popup

UserName

OCHH3COH3COCHCH3OCH3OCHH3COCH3OCHH3COCH3Zr

CAS RN 17501-44-9

IUPAC Name: zirconium(4+) tetrakis(2,4-dioxopentan-3-ide)
Synonyms: zirconium acetylacetonate
Molecular Weight (g/mol): 487.66
Molecular Formula: C20H28O8Zr
InChi Key: DFPGFWYWSAYLCW-UHFFFAOYSA-N
SMILES: [Zr+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)