missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium carbonate, basic hydrate, Thermo Scientific Chemicals
$188.89 - $614.33
Chemical Identifiers
| CAS | 57219-64-4 |
|---|---|
| Molecular Formula | CH2O7Zr2 |
| Molecular Weight (g/mol) | 308.47 |
| MDL Number | MFCD00172265 |
| InChI Key | ACWOHNQUPJAZIT-UHFFFAOYSA-J |
| Synonym | hydrogen carbonate; zirconium 4+, zirconium 4+ ion tetrabicarbonate, zirconium iv carbonatebasic |
| PubChem CID | 22645944 |
| IUPAC Name | [({[hydroxy(oxo)zirconio]oxy}carbonyl)oxy]zirconiumoylol |
| SMILES | O[Zr](=O)OC(=O)O[Zr](O)=O |
Description
Zirconium Carbonate is used in the manufacturing of paint drier auxiliaries, pigments, water proof.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 57219-64-4 | |
| 308.47 | |
| ACWOHNQUPJAZIT-UHFFFAOYSA-J | |
| 22645944 | |
| O[Zr](=O)OC(=O)O[Zr](O)=O |
| CH2O7Zr2 | |
| MFCD00172265 | |
| hydrogen carbonate; zirconium 4+, zirconium 4+ ion tetrabicarbonate, zirconium iv carbonatebasic | |
| [({[hydroxy(oxo)zirconio]oxy}carbonyl)oxy]zirconiumoylol |
Specifications
| White | |
| Powder | |
| CH2O7Zr2 | |
| hydrogen carbonate; zirconium 4+, zirconium 4+ ion tetrabicarbonate, zirconium iv carbonatebasic | |
| ACWOHNQUPJAZIT-UHFFFAOYSA-J | |
| [({[hydroxy(oxo)zirconio]oxy}carbonyl)oxy]zirconiumoylol | |
| 22645944 | |
| 2.2 g/mL |
| 57219-64-4 | |
| 500 g | |
| MFCD00172265 | |
| Insoluble in water. Soluble in hot concentrated acids. | |
| O[Zr](=O)OC(=O)O[Zr](O)=O | |
| 308.47 | |
| 290.45 (Anhydrous) | |
| Zirconium carbonate, Basic hydrate |
Safety and Handling
EINECSNumber : 260-633-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only