missing translation for 'onlineSavingsMsg'
Learn More

Zirconium carbonate, basic hydrate, Thermo Scientific Chemicals

Catalog No. AA43245A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43245A3 2 kg
AA4324536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43245A3 Supplier Thermo Scientific Chemicals Supplier No. 043245A3
Only null left

Zirconium Carbonate is used in the manufacturing of paint drier auxiliaries, pigments, water proof.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57219-64-4
Molecular Formula CH2O7Zr2
Molecular Weight (g/mol) 308.47
MDL Number MFCD00172265
InChI Key ACWOHNQUPJAZIT-UHFFFAOYSA-J
Synonym hydrogen carbonate; zirconium 4+, zirconium 4+ ion tetrabicarbonate, zirconium iv carbonatebasic
PubChem CID 22645944
IUPAC Name [({[hydroxy(oxo)zirconio]oxy}carbonyl)oxy]zirconiumoylol
SMILES O[Zr](=O)OC(=O)O[Zr](O)=O

Specifications

Color White
Physical Form Powder
Quantity 2 kg
Solubility Information Insoluble in water. Soluble in hot concentrated acids.
Formula Weight 290.45 (Anhydrous)
Density 2.2 g/mL
Chemical Name or Material Zirconium carbonate, Basic hydrate
EINECSNumber 260-633-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only