Learn More
Zincon monosodium salt
CAS: 62625-22-3 | C20H16N4NaO6S | 463.42 g/mol
Supplier: Thermo Scientific Chemicals B2425606
Description
Zincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsZincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.
Solubility
Soluble in aqueous alkali. Slightly soluble in water and alcohol. Insoluble in other organic solvents.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Zincon monosodium salt | |
| ∼203°C (decomposition) | |
| MFCD00064385 | |
| 3872699 | |
| IABRINWJUBAIIE-JJECXDOKSA-N | |
| 2-[2-[(Z)-N-[(Z)-(6-oxo-3-sulfocyclohexa-2,4-dien-1-ylidene)amino]-C-phenylcarbonimidoyl]hydrazinyl]benzoic acid;sodium | |
| 131856391 |
| 62625-22-3 | |
| C20H16N4NaO6S | |
| 5 g | |
| Soluble in aqueous alkali. Slightly soluble in water and alcohol. Insoluble in other organic solvents. | |
| C1=CC=C(C=C1)C(=NN=C2C=C(C=CC2=O)S(=O)(=O)O)NNC3=CC=CC=C3C(=O)O.[Na] | |
| 463.42 | |
| 462.42 |
Chemical Identifiers
| 62625-22-3 | |
| 463.42 | |
| IABRINWJUBAIIE-JJECXDOKSA-N | |
| 2-[2-[(Z)-N-[(Z)-(6-oxo-3-sulfocyclohexa-2,4-dien-1-ylidene)amino]-C-phenylcarbonimidoyl]hydrazinyl]benzoic acid;sodium |
| C20H16N4NaO6S | |
| MFCD00064385 | |
| 131856391 | |
| C1=CC=C(C=C1)C(=NN=C2C=C(C=CC2=O)S(=O)(=O)O)NNC3=CC=CC=C3C(=O)O.[Na] |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 263-651-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only