missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Sulfate Monohydrate, Purified, Reagents
Zinc Sulfate Monohydrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
$75.20 - $227.97
Chemical Identifiers
| CAS | 7446-19-7 |
|---|---|
| Molecular Formula | H2O5SZn |
| Molecular Weight (g/mol) | 179.45 |
| InChI Key | RNZCSKGULNFAMC-UHFFFAOYSA-L |
| Synonym | Zinc Sulphate Monohydrate |
| IUPAC Name | zinc(2+) hydrate sulfate |
| SMILES | O.[Zn++].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-336-864
|
Labchem Inc
C235400025A5 |
2.5 kg |
Each for $227.97
|
|
|||||
|
01-336-863
|
Labchem Inc
C2354000500A5 |
500 g |
Each for $75.20
|
|
|||||
Chemical Identifiers
| 7446-19-7 | |
| 179.45 | |
| Zinc Sulphate Monohydrate | |
| O.[Zn++].[O-]S([O-])(=O)=O |
| H2O5SZn | |
| RNZCSKGULNFAMC-UHFFFAOYSA-L | |
| zinc(2+) hydrate sulfate |
Specifications
| Colorless/White | |
| Solid | |
| H2O5SZn | |
| RNZCSKGULNFAMC-UHFFFAOYSA-L | |
| zinc(2+) hydrate sulfate | |
| Pure | |
| 3.2 |
| 7446-19-7 | |
| 2.5 kg | |
| Zinc Sulphate Monohydrate | |
| O.[Zn++].[O-]S([O-])(=O)=O | |
| 179.45 | |
| Poly Bottle | |
| Zinc Sulfate Monohydrate, Purified |