missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate Monohydrate, Purified, Reagents

Catalog No. 01336864
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336864 2.5 kg
01336863 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336864 Supplier Labchem Inc Supplier No. C23540002.5A5
Only null left

Zinc Sulfate Monohydrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7446-19-7
Molecular Formula H2O5SZn
Molecular Weight (g/mol) 179.45
InChI Key RNZCSKGULNFAMC-UHFFFAOYSA-L
Synonym Zinc Sulphate Monohydrate
IUPAC Name zinc(2+) hydrate sulfate
SMILES O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 2.5 kg
Grade Pure
Packaging Poly Bottle
Specific Gravity 3.2
Chemical Name or Material Zinc Sulfate Monohydrate, Purified