missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate, Heptahydrate, Crystal, ACS, 99.0-103.0%, Spectrum™ Chemical

Catalog No. 18614809
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614809 125 g Poly Bottle
18614810 500 g Poly Bottle
18614811 2.5 kg Poly Bottle
18614812 12 kg Poly Pail
18614813 25 kg Fiber Drum
18614814 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18614809 Supplier Spectrum Chemical Mfg Cor Supplier No. Z1095125GM
Only null left

Chemical Identifiers

CAS 7446-20-0
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
IUPAC Name zinc(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

pH 4.4 to 6.0
CAS Max % 1
Quantity 125 g
Percent Purity 99 to 103%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.