missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate, Heptahydrate, ACS, 99.5 to 103.0%, MilliporeSigma™

Supplier:  Technidata 1088831000

 View more versions of this product

Catalog No. M1088831000


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Zinc Sulfate, Heptahydrate
White
Powder
99.5 to 103.0%
ZnSO4 · 7H2O
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
287.54
CHEBI:32312
ACS/EP/ISO/Reagent
EMSURE™, For Analysis
7446-20-0
1 kg
H14O11SZn
MFCD00149894
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc(2+) heptahydrate sulfate
62640
287.54
1.97g/cm3 (20°C); Bulk Density: 800 to 1000 kg/m3

Chemical Identifiers

7446-20-0
287.54
RZLVQBNCHSJZPX-UHFFFAOYSA-L
62640
zinc(2+) heptahydrate sulfate
H14O11SZn
MFCD00149894
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
CHEBI:32312
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store without restrictions

SDS
Documents

Documents

Promotions

Promotions