missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate, Heptahydrate, ACS, 99.5 to 103.0%, MilliporeSigma™

Catalog No. M1088830500
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1088830500 500 g
M1088831000 1 kg
M1088835000 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1088830500 Supplier Technidata Supplier No. 1.08883.0500

Chemical Identifiers

CAS 7446-20-0
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
MDL Number MFCD00149894
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
PubChem CID 62640
ChEBI CHEBI:32312
IUPAC Name zinc(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.5 to 103.0%
Linear Formula ZnSO4 · 7H2O
Formula Weight 287.54
Grade ACS/EP/ISO/Reagent
Density Bulk Density: 800 to 1000 kg/m3
Chemical Name or Material Zinc Sulfate, Heptahydrate, EMSURE™, For Analysis
Recommended Storage Store without restrictions