Learn More
Zinc sulfate heptahydrate, 99%, ACS reagent
CAS: 7446-20-0 | H14O11SZn | 287.54 g/mol
Supplier: Thermo Scientific Chemicals 424600030
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Zinc sulfate heptahydrate | |
| 7446-20-0 | |
| 4.4 to 6.0 (5% soln. at 25°C) | |
| 3 kg | |
| H14O11SZn | |
| MFCD00149894 | |
| zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
| RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
| zinc;sulfate;heptahydrate | |
| 62640 | |
| 287.53 | |
| ACS Reagent |
| 100.0°C | |
| White | |
| Crystals | |
| 99.0 to 103% (Complexometry) | |
| ZnSO4·7H2O | |
| 15, 10359 | |
| Solubility in water: 960g/L. Other solubilities: 1g/2.5mL glycerol, insoluble in alcohol | |
| O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O | |
| 287.54 | |
| CHEBI:32312 | |
| 99% | |
| Plastic bottle |
Chemical Identifiers
| 7446-20-0 | |
| 287.54 | |
| RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
| 62640 | |
| O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
| H14O11SZn | |
| MFCD00149894 | |
| zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
| CHEBI:32312 |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RTECSNumber : ZH5300000
RUO – Research Use Only