missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfate heptahydrate, 98%

Catalog No. p-7020831
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
2500 g
500 g
Catalog No. Quantity
AAA1291536 500 g
AAA129150E 2500 g
2 options

Catalog No. AAA1291536

Supplier: Thermo Scientific Chemicals A1291536

Only null left

CAS: 7446-20-0 | H14O11SZn | 287.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Zinc sulfate heptahydrate is widely used as a coagulant in the production of rayon. It finds an application in the manufacturing of calico printing, wood preservation and lithopone. It is the source of zinc which is supplied in animal feeds, fertilizers and agricultural sprays. It is utilized as electrolytes for zinc plating and surface treating agents. It is also used as a mordant in dyeing, preservative for skin and leather and in medicine as an astringent and emetic.

Solubility
Soluble in water and glycerol.

Notes
Incompatible with strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 7446-20-0
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
MDL Number MFCD00149894
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
PubChem CID 62640
ChEBI CHEBI:32312
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Melting Point 100°C
pH 4.4 to 6
Quantity 500 g
Linear Formula ZnSO4·7H2O
UN Number UN3077
Merck Index 14,10159
Solubility Information Soluble in water and glycerol.
IUPAC Name zinc;sulfate;heptahydrate
Formula Weight 287.54 (161.43 Anhydrous)
Percent Purity 98%
Sensitivity Hygroscopic
Density 1.97
Chemical Name or Material Zinc sulfate heptahydrate
Show More Show Less
Hazard Category H302-H318-H373
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 231-793-3
RTECSNumber ZH5300000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only