missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Sulfate, 0.0200 M (M/50), Ricca Chemical
$95.91 - $638.72
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H14O11SZn |
| Molecular Weight (g/mol) | 287.54 |
| MDL Number | MFCD00149894 |
| InChI Key | RZLVQBNCHSJZPX-UHFFFAOYSA-L |
| Synonym | zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o |
| PubChem CID | 62640 |
| ChEBI | CHEBI:32312 |
| IUPAC Name | zinc(2+) heptahydrate sulfate |
| SMILES | O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R9657000500
|
Ricca Chemical Company
R9657000500A |
500 mL |
Each for $95.91
|
|
|||||
|
R96570001A
|
Ricca Chemical Company
R96570001A |
1 L |
Each for $638.72
|
|
|||||
|
R96570004A
|
Ricca Chemical Company
R96570004A |
4 L |
Each for $225.56
|
|
|||||
Chemical Identifiers
| 7732-18-5 | |
| 287.54 | |
| RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
| 62640 | |
| zinc(2+) heptahydrate sulfate |
| H14O11SZn | |
| MFCD00149894 | |
| zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
| CHEBI:32312 | |
| O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
Specifications
| Colorless | |
| Liquid | |
| H14O11SZn | |
| zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
| O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O | |
| 287.54 | |
| CHEBI:32312 | |
| Natural Poly Bottle |
| 7732-18-5,7446-20-0 | |
| 500 mL | |
| MFCD00149894 | |
| RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
| zinc(2+) heptahydrate sulfate | |
| 62640 | |
| Laboratory | |
| Zinc Sulfate |