Learn More
Zinc Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$301.19 - $602.38
Chemical Identifiers
| CAS | 7440-66-6 |
|---|---|
| Molecular Formula | Zn |
| Molecular Weight (g/mol) | 65.38 |
| MDL Number | MFCD00011291 |
| InChI Key | HCHKCACWOHOZIP-UHFFFAOYSA-N |
| PubChem CID | 23994 |
| ChEBI | CHEBI:30185 |
| IUPAC Name | zinc |
| SMILES | [Zn] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PLZN2-2Y
|
SPEX CertiPrep Group LLC
PLZN22Y |
125 mL | Triple-leached LDPE bottle |
Each for $301.19
|
|
||||
|
PLZN2-2X
|
SPEX CertiPrep Group LLC
PLZN22X |
500 mL | Triple-leached LDPE bottle |
Each for $602.38
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Zn | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7440-66-6 | |
| 65.38 | |
| HCHKCACWOHOZIP-UHFFFAOYSA-N | |
| CHEBI:30185 | |
| [Zn] |
| Zn | |
| MFCD00011291 | |
| 23994 | |
| zinc |
Specifications
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up 30 elements at ppb levels. | |
| 1.0,5.0,94.0 | |
| Zn | |
| AA, ICP | |
| Certified Reference Material | |
| [Zn] | |
| zinc | |
| 125 mL | |
| CHEBI:30185 | |
| Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mL PLZN2-2Y, -2T, -2X 10,000 PLZN2-3Y, -3X 2-5% HCl 1000μg/mL PLZN1-2X 10,000 PLZN1-3X |
| 7440-66-6,7697-37-2,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 1.0,5.0,94.0 | |
| MFCD00011291 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| HCHKCACWOHOZIP-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 65.38 | |
| 23994 | |
| Plasma | |
| Zinc Standard |
Safety and Handling
Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only