missing translation for 'onlineSavingsMsg'
Learn More

Zinc protoporphyrin, 96%

Catalog No. p-7049959
Change view
Click to view available options
Quantity:
10 mg
50 mg
0.25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA32068MA 10 mg
AA32068MF 50 mg
AA3206801 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. AA32068MA missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 032068MA

CAS: 15442-64-5 | C34H32N4O4Zn | 626.034 g/mol

Zinc protoporphyrin-9 is a potent, selective heme oxygenase inhibitor, interleukin-1 antagonist, GCS inhibitor, and NOS1, NOS2, and NOS3 inhibitor. In olfactory neuronal cultures, this product has been used to block CO biosynthesis, which lowered cGMP concentrations. Zinc protoporphyrin-9 is an extremely useful tool to elucidate the role of CO in signal transduction and as a neurotransmitter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15442-64-5
Molecular Formula C34H32N4O4Zn
Molecular Weight (g/mol) 626.034
MDL Number MFCD00011612
InChI Key FUTVBRXUIKZACV-UHFFFAOYSA-L
Synonym zinc protoporphyrin
PubChem CID 131664383
IUPAC Name zinc;3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoate
SMILES CC1=C(C2=CC3=NC(=CC4=C(C(=C(N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C=C)C)C=C)C(=C3CCC(=O)O)C)CCC(=O)[O-].[Zn+2]

Specifications

Color Purple to Red
Odor Odorless
Quantity 10 mg
Solubility Information Insoluble in water. Soluble in ethanol,and DMSO,pyridine,or dimethyl formamide.
Formula Weight 626.04
Percent Purity 96%
Physical Form Crystalline
Chemical Name or Material Zinc protoporphyrin
EINECSNumber 239-455-7
RTECSNumber ZG9210300
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only