accessibility menu, dialog, popup

UserName

CAS RN 15442-64-5

IUPAC Name: zinc protoporphyrin
Synonyms: zinc protoporphyrin
Molecular Weight (g/mol): 626.03
Molecular Formula: C34H32N4O4Zn
InChi Key:
SMILES: [Zn++].Cc1c(CCC(O)=O)c2cc3[n-]c(cc4nc(cc5[n-]c(cc1n2)c(C=C)c5C)c(C=C)c4C)c(C)c3CCC(O)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results