missing translation for 'onlineSavingsMsg'
Learn More

Zinc propionate, 97%

Catalog No. p-7027124
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Catalog No. Quantity
AAB2363618 50 g
AAB2363630 250 g
AAB236360B 1000 g
3 options

Catalog No. AAB2363618

Supplier: Thermo Scientific Chemicals B2363618

Only null left

CAS: 557-28-8 | C6H10O4Zn | 211.52 g/mol

Used to reduce irritation from molds, fungi, and bacteria and as a topical antifungal agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to reduce irritation from molds, fungi, and bacteria and as a topical antifungal agent.

Solubility
Soluble in alcohol, ether, chloroform. Miscible with ethanol; soluble in diethyl ether; slightly soluble in chloroform.

Notes
Hygroscopic, protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 557-28-8
Molecular Formula C6H10O4Zn
Molecular Weight (g/mol) 211.52
MDL Number MFCD00045937
InChI Key XDWXRAYGALQIFG-UHFFFAOYSA-L
Synonym zinc propionate, unii-81pxa87xvc, 81pxa87xvc, propanoic acid, zinc salt 2:1, gyna-plex, propionic acid, zinc salt, zinc dipropionate, propanoic acid, zinc salt, zinc propanoate, propanoic acid, zinc salt, basic
PubChem CID 11189
IUPAC Name zinc;propanoate
SMILES [Zn++].CCC([O-])=O.CCC([O-])=O

Specifications

Quantity 50 g
Sensitivity Hygroscopic
Merck Index 14,10153
Solubility Information Soluble in alcohol,ether,chloroform. Miscible with ethanol; soluble in diethyl ether; slightly soluble in chloroform.
Formula Weight 211.53
Percent Purity 97%
Chemical Name or Material Zinc propionate
Hazard Category H318
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 209-167-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only