accessibility menu, dialog, popup

UserName

CAS RN 557-28-8

IUPAC Name: zinc(2+) dipropanoate
Synonyms: zinc(2+) dipropionate
Molecular Weight (g/mol): 211.52
Molecular Formula: C6H10O4Zn
InChi Key: XDWXRAYGALQIFG-UHFFFAOYSA-L
SMILES: [Zn++].CCC([O-])=O.CCC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results