Learn More
Zinc, plasma standard solution, Specpure™ Zn 1000μg/mL
CAS: 7440-66-6 | Zn | 65.38 g/mol
$97.97 - $323.05
Chemical Identifiers
| CAS | 7440-66-6 |
|---|---|
| Molecular Formula | Zn |
| Molecular Weight (g/mol) | 65.38 |
| MDL Number | MFCD00011291 |
| InChI Key | HCHKCACWOHOZIP-UHFFFAOYSA-N |
| PubChem CID | 23994 |
| ChEBI | CHEBI:30185 |
| IUPAC Name | zinc |
| SMILES | [Zn] |
Description
Zinc, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-66-6 | |
| 65.38 | |
| HCHKCACWOHOZIP-UHFFFAOYSA-N | |
| CHEBI:30185 | |
| [Zn] |
| Zn | |
| MFCD00011291 | |
| 23994 | |
| zinc |
Specifications
| 7440-66-6,7732-18-5 | |
| Liquid | |
| MFCD00011291 | |
| Miscible with water | |
| [Zn] | |
| 65.38 | |
| 23994 | |
| Nitric acid: 5.0%; Zinc: 0.1%; Water: 94.9% |
| 23 hPa (17mm Hg) at 20°C | |
| Zn | |
| Colorless | |
| HCHKCACWOHOZIP-UHFFFAOYSA-N | |
| zinc | |
| 50 mL | |
| CHEBI:30185 | |
| Zinc |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only