missing translation for 'onlineSavingsMsg'
Learn More

Zinc, plasma standard solution, Specpure™ 10μg/mL

Catalog No. AA45269AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45269AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45269AE Supplier Thermo Scientific Chemicals Supplier No. 045269AE

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00011291

Specifications

Physical Form Liquid
Concentration Zn in 2% HNO3
Quantity 100 mL
Grade Specpure
Chemical Name or Material Zinc, plasma standard solution, Zn 10μg/mL
Hazard Category H290-H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only