missing translation for 'onlineSavingsMsg'
Learn More

Zinc Phthalocyanine 95.0+%, TCI America™

Catalog No. P07671G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07671G 1 g
P07675G 5 g
P076725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P07671G Supplier TCI America Supplier No. P07671G

Chemical Identifiers

CAS 14320-04-8
Molecular Formula C32H16N8Zn
Molecular Weight (g/mol) 577.916
MDL Number MFCD00041955
InChI Key PODBBOVVOGJETB-UHFFFAOYSA-N
Synonym zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 %
PubChem CID 2735172
ChEBI CHEBI:51218
SMILES C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2]

Specifications

Color Blue-Purple
Quantity 1 g
Formula Weight 577.92
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Zinc Phthalocyanine