accessibility menu, dialog, popup

UserName

CAS RN 14320-04-8

IUPAC Name: zinc(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide
Synonyms: zinc(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide
Molecular Weight (g/mol): 577.92
Molecular Formula: C32H16N8Zn
InChi Key: PODBBOVVOGJETB-UHFFFAOYSA-N
SMILES: [Zn++].[N-]1C2=NC3=C4C=CC=CC4=C([N-]3)N=C3N=C(N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)