missing translation for 'onlineSavingsMsg'
Learn More

Zinc phosphide, 99.999% (metals basis)

Catalog No. p-7059844
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
0.5 g
10 g
2 g
Catalog No. Quantity
AA4545902 0.5 g
AA4545904 2 g
AA4545909 10 g
3 options

Catalog No. AA4545902

Supplier: Thermo Scientific Chemicals 04545902

Only null left

CAS: 1314-84-7 | Zn3P2

Zinc phosphide has useful applications in photovoltaic cells. It is also employed as a laboratory chemical. It plays an important role in medicine and pharmaceuticals. It acts as a fumigant in grain storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-84-7
Molecular Formula Zn3P2
MDL Number MFCD00049631

Specifications

Boiling Point 1100°C
Melting Point 420°C
Physical Form Pieces
Quantity 0.5 g
UN Number UN1714
Merck Index 14,10152
Solubility Information Insoluble in water and ethanol. Soluble in benzene.
Formula Weight 258.1
Percent Purity 99.999%
Sensitivity Moisture sensitive
Density 4.55 g/mL
Assay (metals basis)
Chemical Name or Material Zinc phosphide
Show More Show Less
Hazard Category H260-H300
Hazard Statement GHS H Statement
H260-H300
In contact with water releases flammable gases which may ignite spontaneously.
Fatal if swallowed.
Precautionary Statement P223-P231+P232-P264b-P270-P280-P301+P310-P330-P335+P334-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ZINC PHOSPHIDE
EINECSNumber 215-244-5
RTECSNumber ZH4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only