missing translation for 'onlineSavingsMsg'
Learn More

Zinc phosphate hydrate, tech.

Catalog No. p-4408055
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
2 kg
500 g
Catalog No. Quantity
AA1158936 500 g
AA11589A3 2 kg
2 options

Catalog No. AA1158936

Supplier: Thermo Scientific Chemicals 01158936

Only null left

CAS: 34807-26-6 | O8P2Zn3 | 386.08 g/mol

Zinc phosphate hydrate is used in dentistry. It is commonly used for luting permanent metal restorations and as a base for dental restorations. Zinc phosphate cement is also used for cementation of inlays, crowns, bridges, and orthodontic appliances and occasionally as a temporary restoration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34807-26-6
Molecular Formula O8P2Zn3
Molecular Weight (g/mol) 386.08
MDL Number MFCD00036282
InChI Key LRXTYHSAJDENHV-UHFFFAOYSA-H
Synonym zinc phosphate, delaphos, trizinc diphosphate, zinc orthophosphate, trizinc phosphate, delaphos 2m, bonderite 40, granodine 80, microphos 90, granodine 16nc
PubChem CID 24519

Specifications

Physical Form Powder
Quantity 500 g
Solubility Information Soluble in acetic acid,dilute mineral acids,ammonia. Insoluble in water
SMILES [Zn++].[Zn++].[Zn++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
IUPAC Name trizinc(2+) diphosphate
Formula Weight 386.05 (Anhydrous)
Grade Technical
Chemical Name or Material Zinc phosphate hydrate
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-944-3
RTECSNumber TD0590000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only