missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc oxide, ≥99% (KT), MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Zinc Oxide |
| CAS | 1314-13-2 |
| Physical Form | Neat |
| Linear Formula | ZnO |
| MDL Number | MFCD00011300 |
| Type | Analytical Standard |
| Molecular Weight (g/mol) | 81.39 |
| Quantity | 100 mg |
| Formula Weight | 81.39 |
| Percent Purity | ≥99% (KT) |
| Show More |