missing translation for 'onlineSavingsMsg'
Learn More

Zinc Nitrate, Crystal, 98.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor Z107550KGBL

 View more versions of this product

Catalog No. 18614782


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10196-18-6
50 kg
Reagent
To Pass Test
1
98.5%
Fiber Drum

Chemical Identifiers

10196-18-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.