missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Nitrate, Crystal, 98.5%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 25 g |
| Percent Purity | 98.5% |
| Grade | Reagent |
| Packaging | Amber Glass Bottle |
| Appearance | To Pass Test |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.