missing translation for 'onlineSavingsMsg'
Learn More

Zinc N-Ethyl-N-phenyldithiocarbamate 98.0+%, TCI America™

Catalog No. p-7128871
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E016325G 25 g
E0163500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E016325G Supplier TCI America Supplier No. E016325G

Chemical Identifiers

CAS 14634-93-6
Molecular Formula C18H20N2S4Zn
Molecular Weight (g/mol) 457.992
MDL Number MFCD00059686
InChI Key KMNUDJAXRXUZQS-UHFFFAOYSA-L
Synonym N-Ethyl-N-phenyldithiocarbamic Acid Zinc Salt
PubChem CID 6093375
IUPAC Name zinc;N-ethyl-N-phenylcarbamodithioate
SMILES CCN(C1=CC=CC=C1)C(=S)[S-].CCN(C1=CC=CC=C1)C(=S)[S-].[Zn+2]

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 25 g
Formula Weight 457.99
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Zinc N-Ethyl-N-phenyldithiocarbamate
EINECSNumber (3)-2186
RTECSNumber ZH0890000
TSCA Yes