missing translation for 'onlineSavingsMsg'
Learn More

Zinc N-Éthyl-N-phényldithiocarbamate 98,0+%, TCI America™

Numéro de catalogue. E016325G
Modifier l'affichage
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E016325G 25 g
E0163500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. E016325G Fournisseur TCI America Code fournisseur E016325G

Chemical Identifiers

CAS 14634-93-6
Molecular Formula C18H20N2S4Zn
Molecular Weight (g/mol) 457.992
MDL Number MFCD00059686
InChI Key KMNUDJAXRXUZQS-UHFFFAOYSA-L
Synonym N-Ethyl-N-phenyldithiocarbamic Acid Zinc Salt
PubChem CID 6093375
IUPAC Name le zinc; N-éthyle-N-phénylcarbamodithioate
SMILES CCN(C1=CC=CC=C1)C(=S)[S-].CCN(C1=CC=CC=C1)C(=S)[S-].[Zn+2]

Specifications

Melting Point 206°C
Color Blanc-jaune
Quantity 25 g
Formula Weight 457.99
Percent Purity ≥98.0% (T)
Physical Form Poudre cristalline
Chemical Name or Material Zinc N-Ethyl-N-phenyldithiocarbamate
EINECSNumber (3)-2186
RTECSNumber ZH0890000
TSCA Yes