missing translation for 'onlineSavingsMsg'
Learn More

Zinc(II) Bis(trifluoromethanesulfinate) Dihydrate 98.0+%, TCI America™

Catalog No. Z00281G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z00281G 1 g
Z00285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z00281G Supplier TCI America Supplier No. Z00281G

Chemical Identifiers

CAS 39971-65-8
Molecular Formula C2F6O4S2Zn
Molecular Weight (g/mol) 331.508
MDL Number MFCD22374174
InChI Key UANOWFITUWBPCF-UHFFFAOYSA-L
Synonym Bis(trifluoromethanesulfinic Acid) Zinc(II) Salt, TFMS
PubChem CID 14174735
IUPAC Name zinc;trifluoromethanesulfinate
SMILES C(F)(F)(F)S(=O)[O-].C(F)(F)(F)S(=O)[O-].[Zn+2]

Specifications

Melting Point 157°C
Color White
Quantity 1 g
Formula Weight 331.51
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Zinc(II) Bis(trifluoromethanesulfinate) Dihydrate
TSCA No
Recommended Storage Refrigerator