Learn More
Epredia™ Zinc Formalin Specimen Transportation System - STS™ Prefilled Specimen Containers
Transport, store and retrieve pathology specimens in a safe and cost effective manner with the Epredia™ Zinc Formalin Specimen Transportation System - STS™ Prefilled Specimen Containers.
$423.53 - $484.27
Specifications
Material Polypropylene
Color Clear
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
22-110-670
View Documents
Epredia™
59401ZF
40 mL, pre-filled 27 mL
Case of 168 Each for $447.10
Only null left
22-110-672
View Documents
Epredia™
59601ZF
60 mL, pre-filled 40 mL
Case of 126 Each for $473.90
Only null left
22-110-673
View Documents
Epredia™
59901ZF
90 mL, pre-filled 60 mL
Case of 120 Each for $452.07
Only null left
22-110-667
View Documents
Epredia™
591201ZF
120 mL, pre-filled 80 mL
Case of 90 Each for $484.27
Only null left
22-110-669
View Documents
Epredia™
59201ZF
20 mL, 14 mL pre-filled
Case of 192 Each for $423.53
Only null left
Description
Description
Closure System
  • Patented Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
Pre-Filled Containers
  • Nonsterile containers marked with a lot code and expiration date
  • Comply with all updated federal and state OSHA requirement
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Specifications
Specifications
Polypropylene
Clear
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'