missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Zinc Formalin Specimen Transportation System - STS™ Prefilled Specimen Containers

Transport, store and retrieve pathology specimens in a safe and cost effective manner with the Epredia™ Zinc Formalin Specimen Transportation System - STS™ Prefilled Specimen Containers.

Supplier:  Epredia™ 59901ZF

 View more versions of this product

Catalog No. 22-110-673


Only null left
Explore available promotions

Description

Description

Closure System
  • Patented Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
Pre-Filled Containers
  • Nonsterile containers marked with a lot code and expiration date
  • Comply with all updated federal and state OSHA requirement
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Specifications

Specifications

Yes
Non-sterile
Bio-Tite Lid
Prefilled Container with 10% Zinc Formalin
Polypropylene
90 mL, pre-filled 60 mL
Clear
120/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions