missing translation for 'onlineSavingsMsg'
Learn More

Zinc Cyanide, Technical, Approx. 85%, Spectrum™ Chemical

Catalog No. 18614744
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614744 25 g Poly Bottle
18614746 500 g Poly Bottle
18614743 2.5 kg Poly Bottle
18614742 12 kg Fiber Drum
18614745 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18614744 Supplier Spectrum Chemical Mfg Cor Supplier No. Z106625GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 557-21-1

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity ∼85%
Grade Technical
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.