missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride, puriss. p.a. ACS Reagent, Honeywell Fluka™

Catalog No. 6001778
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
25 kg
250 g
6 x 250 g
Catalog No. Quantity
6001778 250 g
6001777 1 kg
60045968 6 x 250 g
6001779 25 kg
4 options

Catalog No. 6001778

Supplier: Honeywell Chemicals 31650250G

Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Quantity 250 g
Linear Formula ZnCl2
UN Number 2331
Formula Weight 136.3g/mol
Percent Purity ≥98%
Grade ACS/puriss. p.a.
Vapor Pressure 1mmHg (428°C)
Chemical Name or Material Zinc Chloride, Reag. Ph. Eur. reag. ISO
Hazard Statement P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H302-H314-H410
EINECSNumber 231-592-0
RTECSNumber ZH1400000