missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride, Granular, Technical, Spectrum™ Chemical

$310.73 - $6299.93

Chemical Identifiers

CAS 7646-85-7
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614734
View Documents
Spectrum Chemical Mfg Cor
Z1060500GM
500 g Poly Bottle
Each for $310.73
Only null left
 
18614733
View Documents
Spectrum Chemical Mfg Cor
Z106025KG
2.5 kg Poly Bottle
Each for $1,062.70
Only null left
 
18614732
View Documents
Spectrum Chemical Mfg Cor
Z106012KG
12 kg Poly Pail
Each for $3,246.23
Only null left
 
18614735
View Documents
Spectrum Chemical Mfg Cor
Z106050KGBL
50 kg Fiber Drum
Each for $6,299.93
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7646-85-7

Specifications

7646-85-7
500 g
Poly Bottle
1
Technical
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.