missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride (Cryst./Certified ACS), Fisher Chemical™

Catalog No. p-34599
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
Z33100 100 g Poly Bottle
Z33500 500 g Poly Bottle
Z333 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. Z33100 Supplier Thermo Fisher Scientific Supplier No. Z33100
Only null left

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Boiling Point 732°C
Melting Point 293°C
Color White
pH 5
Physical Form Solid
Quantity 100 g
Assay Percent Range ≥97 %
UN Number 2331
Formula Weight 136.3
Percent Purity ≥97%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 1.3mbar at 428°C
Chemical Name or Material Zinc Chloride
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes burns by all exposure routes. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:1
Instability:1
DOTInformation DOT Class 8, : Corrosive