Learn More
Zinc chloride, anhydrous, 98+%
Zinc chloride, >-98%, Cl2Zn, CAS Number-7646-85-7, zinkchloride, zinc ii chloride, zintrace, zinc dichloride, zinc chloride zncl2, zinc chloride, anhydrous, zine dichloride, zinc chloride fume, zinc chloride, zinc butter, 500g, 732 deg.C, CHEBI:49976, 2.91 | CAS: 7646-85-7 | Cl2Zn | 136.28 g/mol
$55.48 - $1573.11
Chemical Identifiers
CAS | 7646-85-7 |
---|---|
Molecular Formula | Cl2Zn |
Molecular Weight (g/mol) | 136.28 |
MDL Number | MFCD00011295 |
InChI Key | JIAARYAFYJHUJI-UHFFFAOYSA-L |
Synonym | zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride |
PubChem CID | 5727 |
ChEBI | CHEBI:49976 |
IUPAC Name | dichlorozinc |
SMILES | Cl[Zn]Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1628122
|
Thermo Scientific Chemicals
A1628122 |
100 g |
Each for $55.48
|
|
||||||
AAA1628136
|
Thermo Scientific Chemicals
A1628136 |
500 g |
Each for $127.96
|
|
||||||
AAA162810E
|
Thermo Scientific Chemicals
A162810E |
2500 g |
Each for $459.95
|
|
||||||
AAA162810C
|
Thermo Scientific Chemicals
A162810C |
10,000 g |
Each for $1,573.11
|
|
||||||
Description
Zinc chloride is used as a catalyst, dehydrating and condensing agent, soldering flux, and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is likewise employed as a mordant in printing and dyeing materials and in the vulcanizing processes of fiber and rubber. It is useful as an electrolyte in dry cell batteries, in metal industry, in galvanizing iron and as an electrolyte for electroplating. It is useful in organic synthesis, for example, in the Friedel-Crafts acylatoin, Fisher indole synthesis, Lucas reagent (zinc chloride with HCl), activation of allylic/benzylic halides towards reaction with olefins or with sodium cyanoborohydride catalyzed reduction of halides, organozinc reagents for use in Negishi coupling, and aldol condensation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsZinc chloride is used as a catalyst, dehydrating and condensing agent, soldering flux, and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is likewise employed as a mordant in printing and dyeing materials and in the vulcanizing processes of fiber and rubber. It is useful as an electrolyte in dry cell batteries, in metal industry, in galvanizing iron and as an electrolyte for electroplating. It is useful in organic synthesis, for example, in the Friedel-Crafts acylatoin, Fisher indole synthesis, Lucas reagent (zinc chloride with HCl), activation of allylic/benzylic halides towards reaction with olefins or with sodium cyanoborohydride catalyzed reduction of halides, organozinc reagents for use in Negishi coupling, and aldol condensation reactions.
Solubility
Soluble in water, dilute HCl, alcohol, glycerol, acetone and ether.
Notes
Hygroscopic.
Chemical Identifiers
7646-85-7 | |
136.28 | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
5727 | |
dichlorozinc |
Cl2Zn | |
MFCD00011295 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
CHEBI:49976 | |
Cl[Zn]Cl |
Specifications
732°C | |
7646-85-7 | |
Cl2Zn | |
MFCD00011295 | |
14,10132 | |
Soluble in water,dilute HCl,alcohol,glycerol,acetone and ether. | |
Cl[Zn]Cl | |
136.28 | |
CHEBI:49976 | |
≥98% | |
Hygroscopic | |
Zinc chloride, anhydrous |
290°C | |
100 g | |
ZnCl2 | |
UN2331 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
dichlorozinc | |
5727 | |
136.29 | |
Odorless | |
2.91 |
Safety and Handling
GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ZINC CHLORIDE, ANHYDROUS
EINECSNumber : 231-592-0
RTECSNumber : ZH1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only