missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc chloride, 99.99%, (trace metal basis), extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Boiling Point | 732.0°C |
| Melting Point | 293.0°C |
| Color | White |
| Physical Form | Crystalline Powder, Crystals and/or Chunks |
| Quantity | 50 g |
| Packaging | Glass bottle |
| Assay Percent Range | 99.99% |
| Density | .98g/mL |
| Linear Formula | ZnCl2 |
| Fieser | 01,1289; 02,464; 03,338; 05,763; 06,676; 08,536; 09,522; 10,461; 11,602; 12,446; 13,44; 14,177; 15,89; 16,391 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Harmful if swallowed. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsi |
| EINECSNumber | 231-592- |
RUO – Research Use Only