missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride, ≥97%, MP Biomedicals
Specifications
Specifications
| Quantity | 100 g |
| Assay Percent Range | ≥97% |
| Formula Weight | 136.3 |
| Chemical Name or Material | Zinc Chloride |
Safety and Handling
| Recommended Storage | Room Temperature |