Learn More
Zinc chloride, 1M in diethyl ether, Thermo Scientific Chemicals
$142.27 - $142.27
Chemical Identifiers
| CAS | 7646-85-7 |
|---|---|
| Molecular Formula | Cl2Zn |
| Molecular Weight (g/mol) | 136.28 |
| MDL Number | MFCD00011295 |
| InChI Key | JIAARYAFYJHUJI-UHFFFAOYSA-L |
| Synonym | zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride |
| PubChem CID | 5727 |
| ChEBI | CHEBI:49976 |
| IUPAC Name | dichlorozinc |
| SMILES | Cl[Zn]Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7646-85-7 | |
| 136.28 | |
| JIAARYAFYJHUJI-UHFFFAOYSA-L | |
| 5727 | |
| dichlorozinc |
| Cl2Zn | |
| MFCD00011295 | |
| zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
| CHEBI:49976 | |
| Cl[Zn]Cl |
Specifications
| 7646-85-7 | |
| Liquid | |
| Cl2Zn | |
| MFCD00011295 | |
| -40°C (-40°F) | |
| zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
| Cl[Zn]Cl | |
| 136.28 | |
| CHEBI:49976 | |
| 1M in diethyl ether | |
| 0.835 g/mL |
| Colorless | |
| 100 mL | |
| ZnCl2 | |
| UN2924 | |
| 14,10132 | |
| JIAARYAFYJHUJI-UHFFFAOYSA-L | |
| dichlorozinc | |
| 5727 | |
| 136.29 | |
| Hygroscopic | |
| Zinc chloride |
Safety and Handling
GHS H Statement
H225-H314-H302-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P363-P370+P378q-P372-P374-P380-P50
H224-H302-H314-H336
DOTInformation : Transport Hazard Class: 3; Packaging Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only