missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc chloride, 1M in diethyl ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Flash Point | -40°C (-40°F) |
| Packaging | ChemSeal™ bottles |
| Sensitivity | Hygroscopic |
| Density | 0.835g/mL |
| Linear Formula | ZnCl2 |
| UN Number | UN2924 |
| Merck Index | 14,10132 |
| Show More |
Safety and Handling
| Hazard Category | H224-H302-H314-H335-H336 |
| Hazard Statement | GHS H Statement H225-H314-H302-H336 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Harmful if swallowed. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packaging Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only