missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride, 0.7M soln. in THF

Catalog No. p-7031775
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32962AE 100 mL
AAH32962AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32962AE Supplier Thermo Scientific Chemicals Supplier No. H32962AE
Only null left

CAS: 7646-85-7 | Cl2Zn | 136.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc chloride is a widely used as a moderate-strength Lewis acid. In biochemistry research this is used for a source of the zinc ion. It is also be used alone or with phenols for preserving railway ties, fireproofing timber, and with ammonium chloride as a flux for soldering and etching of metals. Zinc chloride is used in the synthesis of the ferrocenylarenes. It is used for the in situ generation of organozincate reagents by reacting with Grignard compounds and also to catalyze chloromethylation of resins for solid phase peptide synthesis.

Solubility
Soluble in water (4320 mg/ml at 25°C), ethanol (slightly), acetone (slightly), ether, and glycerol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Quantity 100 mL
Linear Formula ZnCl2
UN Number UN1993
Solubility Information Soluble in water (4320mg/ml at 25°C),ethanol (slightly),acetone (slightly),ether,and glycerol.
Formula Weight 136.29
Concentration or Composition (by Analyte or Components) 0.7M soln. in THF
Density 0.948
Chemical Name or Material Zinc chloride
Hazard Category H225-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H224-H314-H318-H302
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 231-592-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only