missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc chloride, 0.7M soln. in THF
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Zinc chloride is a widely used as a moderate-strength Lewis acid. In biochemistry research this is used for a source of the zinc ion. It is also be used alone or with phenols for preserving railway ties, fireproofing timber, and with ammonium chloride as a flux for soldering and etching of metals. Zinc chloride is used in the synthesis of the ferrocenylarenes. It is used for the in situ generation of organozincate reagents by reacting with Grignard compounds and also to catalyze chloromethylation of resins for solid phase peptide synthesis.
Solubility
Soluble in water (4320 mg/ml at 25°C), ethanol (slightly), acetone (slightly), ether, and glycerol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
Zinc chloride is a widely used as a moderate-strength Lewis acid. In biochemistry research this is used for a source of the zinc ion. It is also be used alone or with phenols for preserving railway ties, fireproofing timber, and with ammonium chloride as a flux for soldering and etching of metals. Zinc chloride is used in the synthesis of the ferrocenylarenes. It is used for the in situ generation of organozincate reagents by reacting with Grignard compounds and also to catalyze chloromethylation of resins for solid phase peptide synthesis.
Solubility
Soluble in water (4320 mg/ml at 25°C), ethanol (slightly), acetone (slightly), ether, and glycerol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
Specifications
Specifications
| Quantity | 500 mL |
| Density | 0.948 |
| Linear Formula | ZnCl2 |
| UN Number | UN1993 |
| Solubility Information | Soluble in water (4320mg/ml at 25°C),ethanol (slightly),acetone (slightly),ether,and glycerol. |
| Formula Weight | 136.29 |
| Concentration or Composition (by Analyte or Components) | 0.7M soln. in THF |
| Chemical Name or Material | Zinc chloride |
Safety and Handling
| Hazard Category | H225-H302-H314-H335-H336-H351 |
| Hazard Statement | GHS H Statement H224-H314-H318-H302 Extremely flammable liquid and vapor. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 231-592-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only