missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Zinc-Buffered Formalin Prefilled Click Lock Containers
Ready-to-use Buffered Zinc Formalin in leak-resistant, locked containers arranged in convenient cardboard packs
Specifications
| Product Type | Pre-filled Zinc Formalin Container |
|---|---|
| Closure Included | Yes |
| Graduated | Yes |
| Material | Polypropylene |
| Shape | Cylindrical |
Description
- Ready-to-use Buffered Zinc Formalin in leak-resistant, locked containers arranged in convenient cardboard packs
- Recommended for use in IHC, as BFZ preserves more immunoreactivity than standard NBF
- 20 mL vial contains 10 mL of Buffered Zinc Formalin
Specifications
| Pre-filled Zinc Formalin Container | |
| Yes | |
| Cylindrical | |
| Not Autoclavable | |
| 96 |
| Yes | |
| Polypropylene | |
| Non-sterile | |
| Clear |