missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Zinc-Buffered Formalin Prefilled Click Lock Containers
Description
- Ready-to-use Buffered Zinc Formalin in leak-resistant, locked containers arranged in convenient cardboard packs
- Recommended for use in IHC, as BFZ preserves more immunoreactivity than standard NBF
- 20 mL vial contains 10 mL of Buffered Zinc Formalin
Specifications
Specifications
| Product Type | Pre-filled Zinc Formalin Container |
| Closure Included | Yes |
| Graduated | Yes |
| Material | Polypropylene |
| Shape | Cylindrical |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 60 mL |
| Color | Clear |
| No. per Case | 96 |