Learn More
Zinc arsenide, 99.999% (metals basis), Thermo Scientific Chemicals
Used in photo optic applications and dopants. It is also used as a component in the manufacture of semiconductors.
Supplier: Thermo Scientific Chemicals 03566006
Description
Zinc arsenide is used in photo optic applications and dopants. It is also used as a component in the manufacture of semiconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Zinc arsenide | |
| 12006-40-5 | |
| 5 g | |
| MFCD00049629 | |
| zinc arsenide, trizincarsa-1,4-diene, zinc arsenide, pieces trace metals basis | |
| RHKSESDHCKYTHI-UHFFFAOYSA-N | |
| bis(zincio-λ²-arsanyl) zinc | |
| 25147458 | |
| 99.999% | |
| 5.528 |
| 1015°C | |
| Pieces | |
| As2Zn3 | |
| UN1557 | |
| Insoluble in water. Soluble in dilute sulfuric acid. | |
| [Zn].[Zn]=[As].[Zn]=[As] | |
| 345.98 | |
| 345.95 | |
| Glass bottle | |
| 5.528 |
Chemical Identifiers
| 12006-40-5 | |
| 345.98 | |
| RHKSESDHCKYTHI-UHFFFAOYSA-N | |
| 25147458 | |
| [Zn].[Zn]=[As].[Zn]=[As] |
| As2Zn3 | |
| MFCD00049629 | |
| zinc arsenide, trizincarsa-1,4-diene, zinc arsenide, pieces trace metals basis | |
| bis(zincio-λ²-arsanyl) zinc |
Safety and Handling
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 234-486-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only