missing translation for 'onlineSavingsMsg'
Learn More

Zinc arsenide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7076346
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3566006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3566006 Supplier Thermo Scientific Chemicals Supplier No. 03566006
Only null left

Used in photo optic applications and dopants. It is also used as a component in the manufacture of semiconductors.

Zinc arsenide is used in photo optic applications and dopants. It is also used as a component in the manufacture of semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12006-40-5
Molecular Formula As2Zn3
Molecular Weight (g/mol) 345.98
MDL Number MFCD00049629
InChI Key RHKSESDHCKYTHI-UHFFFAOYSA-N
Synonym zinc arsenide, trizincarsa-1,4-diene, zinc arsenide, pieces trace metals basis
PubChem CID 25147458
IUPAC Name bis(zincio-λ²-arsanyl) zinc
SMILES [Zn].[Zn]=[As].[Zn]=[As]

Specifications

Melting Point 1015°C
Physical Form Pieces
Quantity 5 g
UN Number UN1557
Solubility Information Insoluble in water. Soluble in dilute sulfuric acid.
Formula Weight 345.95
Percent Purity 99.999%
Packaging Glass bottle
Specific Gravity 5.528
Density 5.528
Chemical Name or Material Zinc arsenide
Show More Show Less
Hazard Category H301+H331-H350
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 234-486-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only