missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate Solution, 100 g/L, Ricca Chemical

Catalog No. p-3252635
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
944016 500 mL Natural Poly Bottle
944032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 944016 Supplier Ricca Chemical Company Supplier No. 944016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5970-45-6 , 7732-18-5
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.498
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

CAS Min % 87.0, 10.51
CAS Max % 91.0, 10.93
Packaging Natural Poly Bottle
Quantity 500 mL
Concentration or Composition Notes 100g/L
Chemical Name or Material Zinc Acetate Solution