missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate Solution, 100 g/L, Ricca Chemical

Supplier:  Ricca Chemical Company 944032

Quantity 1 L
Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. 944032

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

5970-45-6 , 7732-18-5
219.498
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
zinc;diacetate;dihydrate
C4H10O6Zn
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
2724192
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Zinc Acetate Solution
87.0, 10.51
C4H10O6Zn
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc;diacetate;dihydrate
2724192
1 L
5970-45-6 , 7732-18-5
91.0, 10.93
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
219.498
Natural Poly Bottle
100g/L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents