Learn More
Zinc acetate dihydrate, 97+%
CAS: 5970-45-6 | C4H10O6Zn | 219.498 g/mol
Supplier: Thermo Scientific Chemicals A1290922
Description
Zinc acetate dihydrate is used in the preparation of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis and zinc sulfide nanoparticles coated on silica particles. It is used in dietary and medical applications. It is also used as lozenges for treating the common cold. It is used as a seeding material to grow zinc oxide nanorods on the microfibers of polyethylene terephthalate fabric. In addition, it finds application as wood preservation, production of other zinc salts, ethylene acetate manufacture, and an analytical reagent in various industries. As a plating inhibitor, it is used in nuclear power plant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water and alcohol.
Notes
Incompatible with zinc salts, alkalies-their carbonates, oxalates, phosphates and sulfides.
Specifications
| Zinc acetate dihydrate | |
| 237°C | |
| 6 to 7 | |
| C4H10O6Zn | |
| 100 g | |
| 3732513 | |
| zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn | |
| BEAZKUGSCHFXIQ-UHFFFAOYSA-L | |
| zinc;diacetate;dihydrate | |
| 2724192 | |
| ≥97% |
| 5970-45-6 | |
| 1.735 | |
| Weak, Characteristic | |
| MFCD00066961 | |
| UN3077 | |
| 14,10128 | |
| Soluble in water and alcohol. | |
| CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2] | |
| 219.498 | |
| 219.50 (183.47 Anhydrous) |
Chemical Identifiers
| 5970-45-6 | |
| MFCD00066961 | |
| zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn | |
| zinc;diacetate;dihydrate |
| C4H10O6Zn | |
| BEAZKUGSCHFXIQ-UHFFFAOYSA-L | |
| 2724192 |
Safety and Handling
GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 209-170-2
RTECSNumber : ZG8750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only